Learn More
BINDER™ Divided Inner Glass Door for Series CB, CBF
BINDER™ Divided inner glass door is an option for use with CO2 incubators CB and CBF
1895.00€ - 3825.00€
Specifications
For Use With (Equipment) | BINDER Series CB and CBF |
---|---|
Material | Silicone |
Type | Access Port |
Product Code | Brand | Features | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Features | Price | Quantity & Availability | ||||||
17373683
|
BINDER™
8612-0029 |
Internal door divided into 4 parts |
1895.00€
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
17383683
|
BINDER™
8612-0030 |
Internal door divided into 6 parts |
2840.00€
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||||
17393683
|
BINDER™
8612-0031 |
Internal door divided into 8 parts |
3825.00€
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
Description
- Inner door divided and compartmentation
- Internal door divided into 4, 6 or 8 (depending on model) parts and compartmentation of the interior
Specifications
BINDER Series CB and CBF | |
Silicone | |
Access Port |
Product Title
BINDER™ Divided Inner Glass Door for Series CB, CBF