missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15483848
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 953079-94-2 | C5H10ClNO2 | 151.59 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953079-94-2
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD06801272
InChI Key OYCLYMMIZJWYJG-UHFFFAOYSA-N
Synonym pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
PubChem CID 44890816
IUPAC Name pyrrolidine-3-carboxylic acid;hydrochloride
SMILES C1CNCC1C(=O)O.Cl

Specifications

Melting Point 30°C to 38°C
Quantity 1 g
Flash Point >110°C (230°F)
Formula Weight 151.59
Percent Purity 95%
Chemical Name or Material DL-beta-Proline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.