Learn More
DLX3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97552PCP
This item is not returnable.
View return policy
Description
DLX3 Polyclonal antibody specifically detects DLX3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
DLX3 | |
Produced in rabbits immunized with E. coli-derived Human DLX3 fragment. | |
0.1 mL |
Immunohistochemistry-Paraffin | |
Antigen and protein A Affinity-purified | |
Primary |
Product Content Correction
Product Title
DLX3 Antibody, PerCP, Novus Biologicals™ > 0.1 mL; PerCP