Learn More
Thermo Scientific™ Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand: Thermo Scientific™ AB0784
34.50 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Specifications
Clear | |
8 | |
Strips of 8 Flat Caps | |
Yes |
Polypropylene | |
250/Cs. | |
PCR Plate Cap Strip | |
96-well Plate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
60855402 | Certificate of Compliance | 21/02/2025 | AB0784 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
0Hours
7Minutes
19Seconds
52 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now