Learn More
Thermo Scientific™ Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand: Thermo Scientific™ AB0783
30.16 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Clear | |
8 | |
Strips of 8 Flat Caps | |
Yes |
Polypropylene | |
120/Cs. | |
PCR Plate Cap Strip | |
96-well Plate |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now