Learn More
Thermo Scientific™ Ultra Clear qPCR Caps, strips of 8
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand: Thermo Scientific™ AB0866
55.59 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Highlights
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Specifications
Clear | |
8 | |
Strips of 8 Optical Caps | |
Yes |
Polypropylene | |
120/Cs. | |
PCR Plate Cap Strip | |
96-well Plate |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now