missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Chicken IgY (H+L) Secondary Antibody, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen SA172002
This item is not returnable.
View return policy
Description
SA1-72002 detects Chicken IgY in chicken samples. SA1-72002 has been successfully used in ELISA and western blot procedures. The SA1-72002 immunogen is from purified chicken IgY.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Chicken IgY (H+L) | |
Polyclonal | |
Unconjugated | |
Donkey | |
Affinity chromatography | |
RUO | |
Chicken | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.075% sodium azide; pH 7.2 | |
Purified chicken IgY. | |
500 μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction