missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey Anti-Goat IgG Biotinylated Antibody, R&D Systems™
Donkey Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems BAF109
121.60 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Western Blot.
Specifications
IgG | |
Polyclonal | |
Biotin | |
Immunoglobulin G, ImmunoglobulinG | |
Donkey | |
Affinity Purified | |
RUO | |
3500 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot | |
IGHG1 | |
Goat IgG | |
250 μg | |
Secondary | |
Detects goat IgG heavy and light chains. In Western blot, approximately 5% cross-reactivity with mouse IgG and rabbit IgG is observed. | |
Goat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Donkey Anti-Goat IgG Biotinylated Antibody, R&D Systems™ > 250ug, Biotin
Spot an opportunity for improvement?Share a Content Correction