missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Bio-Techne NBP1-75609
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
IgG (H+L) | |
Polyclonal | |
DyLight 650 | |
Donkey | |
Affinity purified | |
RUO | |
Secondary | |
Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
Flow Cytometry, Immunomicroscopy | |
LYOPH | |
Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
Purified Goat IgG (H&L) | |
1 mg | |
Adaptive Immunity, Immunology | |
Based on IEP, this antibody reacts with heavy gamma chains on goat IgG and light chains on all goat immunoglobulins. This antibody has been pre-adsorbed against human, mouse, rabbit and rat IgG. | |
Goat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Donkey anti-Goat IgG (H+L) Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™ > 1mg; DyLight 650
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction