missing translation for 'onlineSavingsMsg'
Learn More

Donkey anti-Goat IgG (H+L) Secondary Antibody, PE, Invitrogen™

Donkey Polyclonal Secondary Antibody

Brand:  Invitrogen PA129953

Product Code. 11596882

  • 407.00€ / 500µL
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

PA1-29953 detects Goat IgG from goat samples.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Goat IgG (H+L)
Polyclonal
PE
Donkey
Antigen affinity chromatography
RUO
Goat
Liquid
Immunocytochemistry
1.032 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2
Goat IgG whole molecule
500 μL
Secondary
4° C, store in dark, DO NOT FREEZE!
IgG F(ab')2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers