Learn More
Donkey Anti-Mouse IgG NorthernLights™ NL637-conjugated Antibody, R&D Systems™
Donkey Polyclonal Antibody has been used in 9 publications
Brand:  R&D Systems NL008
Product Code. 18105541
  • 229.00€ / 500µL
Description
IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
NorthernLights 637 (NL637)
Immunoglobulin G, ImmunoglobulinG
Donkey
0.5 mL
Secondary
Mouse
IgG
Flow Cytometry, Immunohistochemistry, Immunocytochemistry
1mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry
IGHG1
Affinity Purified
RUO
3500
Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Donkey Anti-Mouse IgG NorthernLights™ NL637-conjugated Antibody, R&D Systems™ > 0.5mL, NL637