missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen SA510042
This item is not returnable.
View return policy
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, human, mouse, pig and rat. For Western blot applications, 5% non-fat dry milk in PBST or TBST is recommended for blocking and incubation of antibodies. BSA is not recommended.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 680 | |
Donkey | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Rabbit IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction