missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rat IgG (H+L) Secondary Antibody, Rhodamine (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Bio-Techne NBP1-73569
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
IgG (H+L) | |
Polyclonal | |
Rhodamine | |
Donkey | |
Affinity Purified | |
RUO | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
LYOPH | |
Western Blot 1:2000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
Rat IgG whole molecule | |
1 mg | |
Adaptive Immunity, Immunology | |
This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, and Sheep Serum Proteins. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Donkey anti-Rat IgG (H+L) Secondary Antibody, Rhodamine (Pre-adsorbed), Novus Biologicals™ > 1mg; Rhodamine
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction