missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey Anti-Sheep IgG (H+L) Fluorescein-conjugated Antibody, R&D Systems™
Donkey Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems F0125
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Flow Cytometry.
Specifications
IgG | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
IGHG1 | |
Sheep IgG | |
100 Tests | |
Secondary | |
Sheep IgG specific IgG was purified by sheep IgG (H+L) affinity chromatography and absorbed against human IgG, and human, mouse, bovine, and rabbit serum to eliminate cross-reactivity. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
Fluorescein | |
Immunoglobulin G, ImmunoglobulinG | |
Donkey | |
Affinity Purified | |
RUO | |
3500 | |
Sheep | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Donkey Anti-Sheep IgG (H+L) Fluorescein-conjugated Antibody, R&D Systems™ > 100 Tests, Fluorescein
Spot an opportunity for improvement?Share a Content Correction