missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey F(ab) anti-Goat IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Donkey Polyclonal Antibody has been used in 1 publication
Brand: Bio-Techne NB120-6523
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
IgG (H+L) | |
Polyclonal | |
Texas Red | |
Donkey | |
Affinity Purified | |
RUO | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
LYOPH | |
Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
Goat IgG whole molecule | |
1 mg | |
Adaptive Immunity, Immunology | |
No reaction was observed against anti-Papain or anti-Donkey IgG F(c). | |
Goat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Donkey F(ab) anti-Goat IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™ > 1mg; Texas Red
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction