Learn More
Hanna Instruments™ Dosing Pump
Brand:  Hanna Instruments™ BL10-2
 View more versions of this product
Product Code. 10274131
  • 480.00€ / Each
Estimated Shipment: 23-04-2025
Specifications
150 mm
45°C
3 bar
180 mm
10 L/hr.
4 kg
Documents
Manuals
Certificates
View accessory products
One moment while we fetch your results.
Product Title
Hanna Instruments™ Dosing Pump > Flow Rate: 10L/hr.