Learn More
Hanna Instruments™ Dosing Pump
Brand:  Hanna Instruments™ BL5-2
 View more versions of this product
Product Code. 10345721
  • 464.00€ / Each
Estimated Shipment: 18-07-2025
Specifications
150 mm
45°C
7 bar
180 mm
5 L/hr.
4 kg
Documents
One moment while we fetch your results.
Product Title
Hanna Instruments™ Dosing Pump > Flow Rate: 5L/hr.