missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxazosin mesylate, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Selective α1-adrenoceptor antagonist (pKi values are 9.0, 8.5 and 8.4 for human α1B, α1A and α1D receptors respectively). Displays antihypertensive activity.
Specifications
Specifications
CAS | 77883-43-3 |
Molecular Formula | C24H29N5O8S |
Quantity | 50 mg |
Synonym | doxazosin mesylate, cardura, carduran, cardular, cardura xl, cardenalin, dedralen, diblocin, doxazomerck, progandol |
InChI Key | VJECBOKJABCYMF-UHFFFAOYSA-N |
SMILES | COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4COC5=CC=CC=C5O4)N)OC.CS(=O)(=O)O |
IUPAC Name | [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone;methanesulfonic acid |
Molecular Weight (g/mol) | 547.583 |
PubChem CID | 62978 |
ChEBI | CHEBI:4709 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'