Learn More
Tocris Bioscience™ DPTN
Potent and selective A3 adenosine receptor antagonist
Brand: Tocris Bioscience™ 7628/10
181.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
DPTN is a potent and selective A3 adenosine receptor antagonist (Ki values are in the range 0.36 - 1.65 nM (hA3AR) and 1.6 - 8.53 nM (rA3AR)). Reduces intrapulmonary pressure when combined with Dexamethasone (Cat. No. 1126).Specifications
N-[4-(3,5-Dimethylphenyl)-5-(4-pyridinyl)-2-thiazolyl]-3-pyridinecarboxamide dihydrochloride | |
10 mg | |
C22H18N4OS.2HCl |
325767-87-1 | |
Adenosine A3 R |