Learn More
Tocris Bioscience™ DPTN
Potent and selective A3 adenosine receptor antagonist
Brand:  Tocris Bioscience™ 7628/10
181.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18718296
  • 231.00€ / 10mg
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
DPTN is a potent and selective A3 adenosine receptor antagonist (Ki values are in the range 0.36 - 1.65 nM (hA3AR) and 1.6 - 8.53 nM (rA3AR)). Reduces intrapulmonary pressure when combined with Dexamethasone (Cat. No. 1126).
Specifications
Specifications
N-[4-(3,5-Dimethylphenyl)-5-(4-pyridinyl)-2-thiazolyl]-3-pyridinecarboxamide dihydrochloride
10 mg
C22H18N4OS.2HCl
325767-87-1
Adenosine A3 R