Learn More
Dräger™ CMS Chips: Butadiene
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406460
Product Code. 12354609
  • 261.00€ / Each
Estimated Shipment: 21-01-2025
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
5 to 30mg/L
90 to 550 sec.
0°C to 40°C
Butadiene CMS Chip
1 to 25
32°F to 104°F