Learn More
Dräger™ CMS Chips: Carbon Dioxide
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406070
 View more versions of this product
Product Code. 10573612
  • 231.00€ / Each
Estimated Shipment: 21-01-2025
Description
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
25 to 140 sec.
0°C to 40°C
Carbon Dioxide CMS Chip
1000 to 25000
32°F to 104°F