Learn More
Dräger™ CMS Chips: Sulfur Dioxide
Suitable for 10 measurements of one gas
235.00€
Product Code | Brand | Type | Measurement Range | Response Time | Humidity Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Measurement Range | Response Time | Humidity Range | Price | Quantity & Availability | ||
12304649
|
Dräger™
6406180 |
Sulfur Dioxide CMS Chip | 5 to 150 | 40 to 360 sec. | 1 to 40mg/L |
235.00€
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
Sulfur Dioxide CMS Chip |