Learn More
Dräger™ Short-Term Detector Tubes: Ethylene Oxide
Detector tubes
183.00€ - 214.00€
Specifications
Components Measured Ethylene Oxide
Humidity Range 3 to 15mg/L
View More Specs
Products 2
Product Code Brand Components Measured Tube Type Measurement Range Response Time Price Quantity & Availability  
Product Code Brand Components Measured Tube Type Measurement Range Response Time Price Quantity & Availability  
10238702
View Documents
Dräger™
6728961
Ethylene Oxide Ethylene oxide 1/a 1 to 15 8 min.
183.00€
Each
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
10594533
View Documents
Dräger™
6728241
Ethylene Oxide Ethylene oxide 25/a 25 to 500 6 min.
214.00€
Each
Estimated Shipment: 07-01-2025
Description
 Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Ethylene Oxide
3 to 15mg/L