Learn More
Multiroir Controlec™ Drawer Separation Insert for Crystal Multiroir 2000
Creates a compartment within a 1/3-width segment in a Crystal Multiroir drawer. Multiroir Controlec™ Drawer Separation Insert for Crystal Multiroir 2000 is a 50mm horizontal cross piece for a 2L drawer that has been fitted with a longtudinal (vertical) column separation piece.
Brand:  Multiroir Controlec™ 11146MS02004C
Additional Details : Weight : 0.80000kg
Product Code. 11886850
  • 4.95€ / Pack of 10
Estimated Shipment: 24-07-2024
Description
Crystal Multiroir are high quality transparent drawer cabinets. They are completely modular and can be combined at will from all sides to save space. Each drawer can be customized with removable partitions to create any size of compartment for partitioned storage.
  • Can be placed on cabinets, shelves or the floor
  • Each unit includes a handle with label holder
  • High transparency Crystal Polystyrene material
  • Shock resistant
  • UV resistant
  • Soap washable
  • Fall arrest stops on each drawer
  • Easy to assemble, tool-free, slide from right to left and from bottom to top
Specifications
Specifications
10/Pack
1/3 Transverse Drawer Separation Insert
2 mm
Transparent
50 mm
Polystyrene
Sectioned storage
50 mm
Crystal Mutiroir 2000 (2 L) Drawer, Transverse