Learn More
Witeg™ Dreschel Gas Washing Bottle with Safety Head
For use with washing and drying gases in the laboratory.
Brand:  Witeg™ 2 403 250
Product Code. 11807830
  • 60.20€ / Each
Estimated Shipment: 03-02-2025
Description
  • Made from borosilicate glass
  • Acc. to DIN 12596
  • Complete package
Specifications
Specifications
250 mL
With safety head, Without sintered disc
29/32