Learn More
Witeg™ Dreschel Gas Washing Bottle with Safety Head
For use with washing and drying gases in the laboratory.
Brand: Witeg™ 2 403 250
This item is not returnable.
View return policy
Description
- Made from borosilicate glass
- Acc. to DIN 12596
- Complete package
Specifications
250 mL | |
With safety head, Without sintered disc | |
29/32 |