Learn More
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone
Excellent for protecting reactions against moisture while equalizing pressure. Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
23.00€ - 31.00€
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
10007730
|
Lenz Laborglasinstrumente™
05337001 |
NS 14/23 |
23.00€
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
10189600
|
Lenz Laborglasinstrumente™
05337004 |
NS 29/32 |
31.00€
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
10366261
|
Lenz Laborglasinstrumente™
05337002 |
NS 19/26 |
27.00€
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
Description
- Tube bent at a 75° angle
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Drying Tube | |
Bent |
Product Title
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone