missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSCAM Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
383.00€ - 611.00€
Specifications
Antigen | DSCAM |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18336692
|
Bio-Techne
NBP3-17835-25UL |
25 μg |
383.00€
25µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18392605
|
Bio-Techne
NBP3-17835-100UL |
100 μg |
611.00€
100µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
DSCAM Polyclonal antibody specifically detects DSCAM in Human samples. It is validated for ImmunofluorescenceSpecifications
DSCAM | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
CHD2-42, CHD2-52, Down syndrome cell adhesion molecule, human CHD2-52 down syndrome cell adhesion molecule, 10CHD2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: QDGGRVMNMAVPKAHRPGDLIHLPPYLRMDFLLNRGGPGTSRDLSLGQACLEPQKSRTLK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
1826 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title