Learn More
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled are manufactured from durable DURAN™ borosilicate glass.
24.00€ - 74.00€
Specifications
Material Borosilicate Glass, Duran Glass
Bottom Shape Round
View More Specs
Products 14
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
10602662
View Documents
Lenz Laborglasinstrumente™
03213270
1000 mL 19/26 NS Side Neck, 24/29 NS Center Neck
74.00€
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
12633790
View Documents
Lenz Laborglasinstrumente™
03211137
100 mL 14/23 NS Side Neck, 14/23 NS Center Neck
32.00€
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10332611
View Documents
Lenz Laborglasinstrumente™
03214137
100 mL 14/23 NS Side Neck, 29/32 NS Center Neck
24.00€
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10070281
View Documents
Lenz Laborglasinstrumente™
03214237
100 mL 19/26 NS Side Neck, 29/32 NS Center Neck
49.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
10372751
View Documents
Lenz Laborglasinstrumente™
03214149
250 mL 14/23 NS Side Neck, 29/32 NS Center Neck
34.00€
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
10204481
View Documents
Lenz Laborglasinstrumente™
03214249
250 mL 19/26 NS Side Neck, 29/32 NS Center Neck
58.00€
Each
In Stock
to see stock.
Edge
Add to basket
10481083
View Documents
Lenz Laborglasinstrumente™
03213149
250 mL 14/23 NS Side Neck, 24/29 NS Center Neck
51.00€
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
10284521
View Documents
Lenz Laborglasinstrumente™
03214449
250 mL 29/32 NS Side Neck, 29/32 NS Center Neck
40.00€
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
10382751
View Documents
Lenz Laborglasinstrumente™
03214158
500 mL 14/23 NS Side Neck, 29/32 NS Center Neck
40.00€
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
10388839
View Documents
Lenz Laborglasinstrumente™
03214128
50 mL NS29/NS14
41.00€
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
10139120
View Documents
Lenz Laborglasinstrumente™
03214458
500 mL 29/32 NS Side Neck, 29/32 NS Center Neck
46.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
10772512
View Documents
Lenz Laborglasinstrumente™
03211122
24 mL 14/23 NS Side Neck, 14/23 NS Center Neck
25.00€
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
10624271
View Documents
Lenz Laborglasinstrumente™
03211128
50 mL 14/23 NS Side Neck, 14/23 NS Center Neck
24.00€
Each
Estimated Shipment: 02-06-2025
to see stock.
Add to basket
10181991
View Documents
Lenz Laborglasinstrumente™
03214258
500 mL 19/26 NS Side Neck, 29/32 NS Center Neck
69.00€
Each
Estimated Shipment: 23-06-2025
Description
  • Acc. to DIN 12394
  • With side neck at 20° angle
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • Ground joint features high taper accuracy and excellent tightness
Specifications
Specifications
Borosilicate Glass, Duran Glass
Round
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled