Learn More
DWK Life Sciences DURAN™ Slit sieve
For use with paper or membrane filters.
Brand:  DWK Life Sciences 213404404
 View more versions of this product
Product Code. 15251689
  • 413.00€ / Pack of 5
Estimated Shipment: 07-07-2025
Description
DURAN™ slotted screen plates serve as support surfaces for the actual filter (e.g. paper or membrane filter), e.g. in the case of buechner funnels.
  • As support for paper or membrane filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3)
Suitable for coarse filtration without additional filter medium; Filtration
5
5 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Slit sieve > Diameter 95 mm