Learn More
DWK Life Sciences DURAN™ Volumetric Flask, Class A, batch certificate
DURAN™ volumetric flasks are used for the accurate measurement of specific quantities of liquids. They are volumetric analysis aids in quantitative laboratory work. These volumetric flasks are certified by batch.
39.65€ - 171.00€
Specifications
Material | Borosilicate Glass (3.3), Polyethylene |
---|---|
For Use With (Application) | Preparation and storage of standard solutions; Measuring liquid amounts; Diluting solutions; Mixing several components; Measurement analysis; Volumetric measurement; To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250°C in drying cabinets or sterilisers |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15210866
|
DWK Life Sciences
246780159 |
1 mL |
71.05€
Pack of 2 |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||||
15220866
|
DWK Life Sciences
246780253 |
2 mL |
71.05€
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
15230866
|
DWK Life Sciences
246780956 |
5 mL (Wide neck) |
39.65€
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
15287339
|
DWK Life Sciences
246781052 |
10 mL (Wide neck) |
39.90€
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
15250866
|
DWK Life Sciences
246781258 |
20 mL |
41.10€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15260866
|
DWK Life Sciences
246781455 |
25 mL |
41.10€
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
15270866
|
DWK Life Sciences
246781755 |
50 mL |
44.85€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15280866
|
DWK Life Sciences
246782451 |
100 mL (Stopper Size 12/21) |
49.95€
Pack of 2 |
Estimated Shipment: 14-10-2025
Log in to see stock. |
||||||
15290866
|
DWK Life Sciences
246782554 |
100 mL (Stopper Size 14/23) |
50.50€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15200876
|
DWK Life Sciences
246783259 |
200 mL |
57.25€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15210876
|
DWK Life Sciences
246783653 |
250 mL |
59.35€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15220876
|
DWK Life Sciences
246784452 |
500 mL |
70.25€
Pack of 2 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
15230876
|
DWK Life Sciences
246785457 |
1000 mL |
125.00€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15240876
|
DWK Life Sciences
246785551 |
1000 mL (Wide neck) |
137.00€
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15250876
|
DWK Life Sciences
246786359 |
2000 mL |
168.00€
Pack of 2 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15260876
|
DWK Life Sciences
246787355 |
5000 mL |
171.00€
Each |
In Stock
Log in to see stock. |
||||||
Description
Volumetric flasks are available in two different accuracy Classes: Class A and Class B. The two Classes differ in the accuracy of measurement with Class A being the highest accuracy, and Class B is approximately half that of Class A. The volumetric flasks with accuracy Class A have a blue print for clear glass products and a white print for amber versions. Volumetric flasks with a batch number and accuracy Class A are supplied with a batch certificate. This certificate documents the mean value obtained from measuring the batch in question, the standard deviation and the day of issue. The batch certificates can also be retrieved online. The batch number consists of four digits. The first two numbers specify the production year, and the following two numbers specify the batch, e.g.: 18.01.
- Calibration based on the poured in volume (In) at a + 20°C reference temperature
- Outstanding precision during volumetric measurement and high-quality process stability
- Thanks to the exact processing and precisely calibrated scales, they permit the highly accurate determination and measurements of volume
- Supplied with ergonomic polyethylene stopper for easy and safe tightening or removal
- The volume content tolerances conform to accuracy Class A
- Fired-on, highly durable blue print
- With printed ring mark for better visibility
- Influence of undesirable factors is kept to an absolute minimum
Specifications
Borosilicate Glass (3.3), Polyethylene | |
Preparation and storage of standard solutions; Measuring liquid amounts; Diluting solutions; Mixing several components; Measurement analysis; Volumetric measurement; To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250°C in drying cabinets or sterilisers | |
Autoclavable |