Learn More
Eppendorf™ DWP Centrifuge Bucket
Brand: Eppendorf™ 5820743000
1561.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Rotor S-4-104 | |
4/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
91Hours
18Minutes
34Seconds
34New Offer Save Up to 85%
Product Content Correction
Product Title
Eppendorf™ DWP Centrifuge Bucket > For use with: Rotor S-4-104; Quantity: 4/Pk.