Learn More
Fisherbrand™ Dycem™ Polymeric Compound Non-slip Mats
Provides a secure surface that anchors items such as cups, plates, tools, and a variety of other objects.
5.15€ - 11.25€
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
12846445
|
Fisherbrand™
2047/6 |
200 x 200 mm |
5.15€
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
12856445
|
Fisherbrand™
2047/9 |
320 x 260 mm |
11.25€
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
Description
- Can be washed with soap and water
- Latex-free
Specifications
Blue | |
Dychem Polymeric Compound |
Product Title
Fisherbrand™ Dycem™ Polymeric Compound Non-slip Mats