missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 405 NHS Ester

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.

Brand:  Thermo Scientific™ 46401


 View more versions of this product

Product Code. 11839370

  • 299.00€ / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.

The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.

Highlights:

  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate

  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)

  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 x 50 μg
DMF (Dimethylformamide)
Amine
DyLight™ 405
DyLight™
400
Dye
Store desiccated at -20°C, protected from light.
Active Ester, Succinimidyl Ester, NHS Ester
DyLight Fluors
420
Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI4461002 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number46401
Lot NumberZH4423731 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number46401
Lot NumberZG4383522 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog Number46401
Lot NumberZG4348591 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number46401
Lot NumberZF4316912 Certificate TypeCertificate of Analysis Date29/05/2024 Catalog Number46401
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DyLight™ 405 NHS Ester > 405 NHS Ester; 5 x 50μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.