Learn More
Thermo Scientific™ DyLight™ 800-4xPEG NHS Ester
Fluorescently label antibodies and other proteins for near-IR detection with this derivative of our high-performance DyLight 800 Dye.
Brand: Thermo Scientific™ 46604
367.20 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Description
Fluorescently label antibodies and other proteins for near-IR detection with Thermo Scientific™ DyLight™ 800-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 800 Dye.
Thermo Scientific DyLight 800-4xPEG Amine-Reactive Dye is a derivative of our high-performance DyLight 800 Dye that can be used to fluorescently label antibodies and other proteins for near-IR detection.
The DyLight 800-4xPEG dye contains 4 polyethylene glycol (PEG) chains that are non-cytotoxic, enhance fluorescence and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 800-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The NIR fluorescence properties of DyLight 800-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.
Highlights:- High fluorescence intensity – fluorescence comparable to Alexa Fluor™ 800 and IRDye™ 800
- PEGylated – improves solubility in aqueous solution and aids in cell permeability
- Fluorescence microscopy
- In vivo or ex vivo imaging
- Cell-based assays
- Flow cytometry/fluorescence-activated cell sorting (FACS)
DyLight 800-4xPEG Amine-Reactive Dye is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds. Learn more about NHS ester chemistry.
Typical labeling reactions require the dye to first be dissolved in anhydrous dimethyl formamide (DMF) or another suitable organic solvent before adding a specific molar amount of dye to an amine-free buffer containing the protein to be labeled. However, the high solubility of DyLight Fluors permits protein solutions to be added directly to the labeling reagent.

Specifications
1 mg | |
DMF (Dimethylformamide) | |
Amine | |
DyLight™ 800 | |
DyLight™ | |
777 |
Reactive Dye | |
Store desiccated at -20°C. | |
Active Ester, Succinimidyl Ester, NHS Ester | |
DyLight Fluors | |
794 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZH4423734 | Certificate of Analysis | 20/08/2024 | 46604 |
ZA4166857 | Certificate of Analysis | 25/01/2024 | 46604 |
YL4143557 | Certificate of Analysis | 25/01/2024 | 46604 |
YK4113056 | Certificate of Analysis | 07/12/2023 | 46604 |
YJ4091165 | Certificate of Analysis | 02/11/2023 | 46604 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.