missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 594 Maleimide

Product Code. 10118693 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10118693

Brand: Thermo Scientific™ 46608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of maleimide-activated DyLight Dyes.

Thermo Scientific™ Sulfhydryl-Reactive DyLight Dyes are maleimide-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as probes for cellular imaging and other fluorescence detection methods.

The Thermo Scientific™ DyLight™ Dyes have absorption spectra ranging from 350 to 770nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor™ or Cy™ Dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight Reagents allows a high fluor-to-protein ratio without precipitation during conjugation.
The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.

Highlights:
  • Specific — maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
  • Efficient labeling methods — well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
  • Optimized antibody labeling procedure —complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
  • Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
  • Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies
TRUSTED_SUSTAINABILITY

Specifications

Description DyLight 594 Maleimide
Quantity 1 mg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Label or Dye DyLight™ 594
Label Type DyLight Fluors
Product Line DyLight
Emission 618
Excitation 593
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.