Learn More
Thermo Scientific™ DyLight™ 680 Maleimide
Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of maleimide-activated DyLight Dyes.
Brand: Thermo Scientific™ 46618
325.60 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Description
Thermo Scientific™ Sulfhydryl-Reactive DyLight Dyes are maleimide-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as probes for cellular imaging and other fluorescence detection methods.
The Thermo Scientific™ DyLight™ Dyes have absorption spectra ranging from 350 to 770nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor™ or Cy™ Dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight Reagents allows a high fluor-to-protein ratio without precipitation during conjugation.The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.
Highlights:
- Specific — maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
- Efficient labeling methods — well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
- Optimized antibody labeling procedure —complete protocol for IgG reduction and labeling and calculating the labeling efficiency
Applications:
- Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
- Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

Specifications
DyLight 680 Maleimide | |
Maleimide | |
Store desiccated at -20°C. | |
Maleimide | |
DyLight Fluors | |
712 |
1 mg | |
DMF (Dimethylformamide) | |
Thiol | |
DyLight™ 680 | |
DyLight™ | |
692 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZK4499681 | Certificate of Analysis | 29/10/2024 | 46618 |
Z4209952 | Certificate of Analysis | 21/06/2024 | 46618 |
ZA209552 | Certificate of Analysis | 16/02/2024 | 46618 |
YJ4066463 | Certificate of Analysis | 18/10/2023 | 46618 |
YI4015454 | Certificate of Analysis | 23/08/2023 | 46618 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.