missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ MAX EPEC/VTEC O45 Kit

Dynabeads MAX EPEC/VTEC O45 Kit

697.00€

Specifications

Content And Storage Store in refrigerator (2–8°C). DO NOT FREEZE.
Product Type EPEC/VTEC Kit
For Use With (Equipment) Dynabeads™ MPC™, KingFisher mL Food Protection Purification System
Product Line Dynabeads™
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17640466
View Documents
Applied Biosystems™
A14631
100 Reactions
697.00€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Dynabeads™ MAX EPEC/VTEC O45 Kit allows for the rapid, selective concentration of E. coli O45 serogroup bacteria in food and environmental samples to improve downstream detection via PCR or culture methods.

  • Improve detection sensitivity in PCR sample analysis
  • Reduce background in selective agar detection
  • Perform the method manually or automate using the BeadRetriever™ System

    Improve Sensitivity and Reduce Background
    Highly specific antibodies attached to paramagnetic beads are incubated with an aliquot of pre-enriched sample. The beads bind and concentrate the target bacteria from non-target organisms. Washing steps help limit the carryover of inhibitors and matrix-associated particulates. These steps combine to improve the sensitivity of PCR-based detection while reducing background for plating methods.

    Manual or Automated Processing
    The immunomagnetic separation (IMS) steps can be performed manually with magnets and mixers or automated using the BeadRetriever™ System.

    Sensitivity
    Following the Dynabeads™ MAX EPEC/VTEC O45 protocol, the presence or absence of one viable E. coli O45 can be determined in the sample sizes described if this one E. coli cell is able to replicate and is not masked by resident background flora.
  • Specifications

    Specifications

    Store in refrigerator (2–8°C). DO NOT FREEZE.
    EPEC/VTEC Kit
    Dynabeads™ MPC™, KingFisher mL Food Protection Purification System
    Dynabeads™
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Testing of Food and Environmental Samples Only.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Applied Biosystems™ Dynabeads™ MAX EPEC/VTEC O45 Kit

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.