Learn More
3M™ E-A-R™ Express™ Ear Plugs, 28 dB, Corded Product Code.: 15729047
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
3M™ E-A-R™ Express™ Ear Plugs, 28 dB, Corded
Product Code. 15729047
Yellow, Blue
Click to view available options
Color:
Yellow, Blue
Product Code. 15729047
Brand: 3M™ 7000103732
Estimated Shipment: 12-06-2025
3M™ E-A-R™ Express™ Ear Plugs are our recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can reduce noise levels by 28 dB and have a hygienic insertion grip.
  • Reduces noise levels by 28 dB CE certified EN 353
  • Easy to slide gently into your ears with slow recovery patented foam
  • Foam tip gives a comfortable and effective seal
  • Features a hygienic insertion grip
  • Recognisable blue stem and yellow tip for easy identification
  • Cord attachment – convenient if you move in and out of noisy environments
  • 100 pairs per box
  • Available with probed feature for use with 3M™ E-A-RFit™ Dual-Ear Validation System
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, Blue
Certifications/Compliance CE Approved
Product Type Express Pod Earplug
Material Polyurethane
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Express™ Ear Plugs, 28 dB, Corded >