missing translation for 'onlineSavingsMsg'
Learn More

Ebola Virus VP40 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA5117406

Product Code. 17259903

  • 477.00€ / 100µL
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Centrifuge briefly prior to opening.

Ebola virus is an enveloped, negative-stranded RNA virus. This virus belongs to the family Filoviridae and has a distinctive morphology similar only to that of Marburg virus, but apparently has no antigenic characteristics of Marburg virus. Its genome is a nonsegmented RNA strand. SDS-PAGE profiles demonstrate the presence of seven structural proteins. These are the major nucleoprotein NP (104 K), glycoprotein GP (125K), L protein (180 K), and four proteins: VP40 (40 K), ViPX (35 K), VP30 (30 K) and VP24 (24 K). All the proteins are encoded by monocistronic mRNA transcripts complementary to virion RNA. GP is the major protein of the surface spikes and only this 125K viral protein is glycosylated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ebola Virus VP40
Polyclonal
Unconjugated
EVD
Recombinant protein encompassing a sequence within the center region of Ebola virus VP40 Protein. The exact sequence is proprietary.
100 μL
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Western Blot
0.67 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ebola Virus VP40 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.