Learn More
Macherey Nagel EC HPLC column EC 100/4NUCLEODUR 100-5 C8 ec length: 100 mm,
Brand:  Macherey Nagel 760704.40
EC HPLC column EC 100/4NUCLEODUR 100-5 C8 ec length: 100 mm,
Product Code. 12763491
  • 387.00€ / Each
In Stock