Learn More
Fisherbrand™ Plain Economy PTFE Stir Bar
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
Brand:  Fisherbrand™ 001.2912
 View more versions of this product
Product Code. 11775134
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications
Specifications
Autoclavable
White
0.125 in.
0.5 in.
3 mm
PTFE
Stir Bar
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
Containers & Vessels / Stirrer Hot Plates
PTFE
12 mm
10/Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Plain Economy PTFE Stir Bar > Dimensions (Dia. x L): 3 dia.x 12mmL