Learn More
Kolmi Hopen™ Elasticated Helmet Hood with Small Opening
Click to view available options
Elasticated Helmet Hood with Small Opening; Color: White; Design Type: Helmet; Disposable: Yes; Includes: Caps and Polybags; Material: Polypropylene
Description
- Ultra-effective protection for the wearer and the environment
- Available for color code tracking
- 25 g/m2 fabric weight
Specifications
Specifications
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Design Type | Helmet |
Quantity | 50 Pk. |
Includes | Caps and Polybags |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Kolmi Hopen™ Elasticated Helmet Hood with Small Opening >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu