Learn More
WTW™ Electrolyte for WTW™ Amperometric Oxygen Sensors
Brand:  WTW™ 205210
98.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11755104
  • 104.00€ / 50mL
Estimated Shipment: 03-04-2025
Specifications
Electrolyte
10
Amperometric oxygen sensor
Documents
One moment while we fetch your results.
Product Title
WTW™ Electrolyte for WTW™ Amperometric Oxygen Sensors > Capacity: 50mL; pH Range: 10; Physical State: Liquid