Learn More
Thermo Scientific™ ELISA Systems™ Food Allergen Swab Kit for the use with ELISA Systems™ Food Allergen Test Kits.
Simplify food allergen testing in food and environmental samples with this swab kit.
Brand:  Thermo Scientific™ ESSWB-10
74.63 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13285369
  • 75.85€ / Pack of 10
Description
Simplify food allergen testing in food and environmental samples with ELISA Systems™ Food Allergen Swab Kit. The kit comes with sterile swabs and specially formulated, sterile swab wetting solution for each swab.
The monitoring of food allergens in production facilities involves several key stages in accordance with HACCP principles. The use of swab samples can be a valuable tool.
When used appropriately, they can provide detailed information regarding the presence of detectable allergenic material. This is of particular importance in environmental monitoring situations, such as:
  • Verification of sanitation procedures
  • Raw material storage facilities
  • Food preparation facilities
The ELISA Systems Swab Residue Kit is designed to provide a convenient supply of swab items for use in the collection of swab samples.
Not all products are available for sale in all territories. Please inquire.
ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.
ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA Systems Food Allergen Swab Kit
Food Allergen Swab Kit
10 Pk.
ELISA (Standard)
10 x sterile swab wetting solution, 10 x sterile swabs
20°C to 25°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ ELISA Systems™ Food Allergen Swab Kit for the use with ELISA Systems™ Food Allergen Test Kits. >