Learn More
Merck Accessories for XX2702550 1225 Sampling Manifold
Accessories for filter holders for sample preparation.
Brand: Merck XX2702555
280.93 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Accessories for filter holders for sample preparation.Specifications
Not Autoclavable | |
EMD Millipore 1225 sampling vacuum manifold | |
Laboratory Filter Holders or Cyclones |
Sample Cup Extension Barrel | |
Non-sterile | |
59 mL |