Learn More
Rogo Sampaic™ Empty Dissection kit
Brand:  Rogo Sampaic™ FMQ006
Product Code. 11758551
  • 10.00€ / Each
Estimated Shipment: 16-06-2025
Specifications
28 cm
1 Ea.
Empty Dissection Kit
Product Title
Rogo Sampaic™ Empty Dissection kit > Length: 28cm