missing translation for 'onlineSavingsMsg'
Learn More

Endothelial Lipase Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB400-111MFV450

Product Code. 18723193

  • 579.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Endothelial Lipase Polyclonal antibody specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endothelial Lipase
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen
EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
A C-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
0.1 mL
Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Microautophagy, Unfolded Protein Response
9388
Store at 4C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen)
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse (Negative)
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Endothelial Lipase Antibody, mFluor Violet 450 SE, Novus Biologicals™ > 0.1 mL, mFluor Violet 450 SE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.