missing translation for 'onlineSavingsMsg'
Learn More

Endothelial Lipase Antibody (OTI1D9), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-71309MFV610

Product Code. 18484328

  • 795.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Endothelial Lipase Monoclonal antibody specifically detects Endothelial Lipase in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endothelial Lipase
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Immunogen affinity purified
RUO
Primary
Human
Purified
Flow Cytometry
OTI1D9
Flow Cytometry
EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Full length human recombinant protein of human LIPG (NP_006024) produced in HEK293T cell.
0.1 mL
Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Microautophagy, Unfolded Protein Response
9388
Store at 4C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Endothelial Lipase Antibody (OTI1D9), mFluor Violet 610 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 610 SE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.