Learn More
Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
2485.00€ - 4665.00€
Specifications
Connection Type | D-sub connector, 9-pin; USB UTL; USB Device |
---|---|
Display Type | Intuitive Graphic Touch Technology |
Material | Glass, PBT, Stainless Steel |
Model | Electronic Precision Balance |
Voltage | 100/240 V (± 10%) |
Product Code | Brand | Capacity | Readability | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Description | Price | Quantity & Availability | |||
16695643
|
Sartorius
BCA223I-1CEU |
220 g | 1 mg | Verified |
2780.00€
Each |
In Stock
Log in to see stock. |
|||
16615663
|
Sartorius
BCA323I-1CEU |
320 g | 1 mg | Verified |
2975.00€
Each |
In Stock
Log in to see stock. |
|||
16625673
|
Sartorius
BCA423I-1CEU |
420 g | 1 mg | Verified |
3180.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16645683
|
Sartorius
BCA623I-1CEU |
620 g | 1 mg | Verified |
3570.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16665693
|
Sartorius
BCA822I-1CEU |
820 g | 10 mg | Verified |
2485.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16675633
|
Sartorius
BCA1203I-1CEU |
1200 g | 1 mg | Verified |
4665.00€
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||
16655633
|
Sartorius
BCA1202I-1CEU |
1200 g | 10 mg | Verified |
2585.00€
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
16646243
|
Sartorius
BCA2202I-1CEU |
2200 g | 10 mg | Verified |
2780.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16635643
|
Sartorius
BCA2201I-1CEU |
2200 g | 100 mg | Verified |
2485.00€
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
16696243
|
Sartorius
BCA3202I-1CEU |
3200 g | 10 mg | Verified |
2870.00€
Each |
In Stock
Log in to see stock. |
|||
16685663
|
Sartorius
BCA4202I-1CEU |
4200 g | 10 mg | Verified |
2975.00€
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||
16665673
|
Sartorius
BCA5201I-1CEU |
5200 g | 100 mg | Verified |
2680.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16605683
|
Sartorius
BCA6202I-1CEU |
6200 g | 10 mg | Verified |
3570.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16625693
|
Sartorius
BCA8201I-1CEU |
8200 g | 100 mg | Verified |
2780.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16606243
|
Sartorius
BCA10201I-1CEU |
10,200 g | 100 mg | Verified |
3570.00€
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
16695633
|
Sartorius
BCA12201I-1CEU |
12,200 g | 100 mg | Verified |
3865.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
Description
- Entris™ II Advanced Balances - Quality, Value, and Consistency
- For accurate weighing results that can be trusted time after time
- Simplifies balance leveling with built-in, real-time level support
- Increases reliability of weighing results using integrated protection systems
- Controlled access to balance settings with user management
- Gapless documentation thanks to CalAuditTrail
Specifications
D-sub connector, 9-pin; USB UTL; USB Device | |
Glass, PBT, Stainless Steel | |
100/240 V (± 10%) | |
50/60 Hz |
Intuitive Graphic Touch Technology | |
Electronic Precision Balance | |
0.2 A |