Learn More
Memmert™ Entry Port 14 mm for UF110
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF110D6
Additional Details : Weight : 0.01200kg
Description
- Entry port with 14 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
UF110 Oven | |
Entry Port | |
14 mm |